Disodium sulphide nonahydrate ≥98.0% ACS
Catalog # 36622.30
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:250 g
- Seq:3545
- Environmentally Preferable:
Specifications
About this item
Synonyms: Sodium sulphide nonahydrate
CAS Number:1313-84-4
:
Formula:Na₂S·9H₂O
MDL Number:MFCD00149184
:240.18 g/mol
Boiling Pt:920
Melting Pt:∼ 50 °C (dec.)
Density:1.427
EINECS:215-211-5
UN:1849
REACH:01-2119513694-38
:GHS06
:GHS05
:GHS09
Storage Temperature:Ambient
ADR:8 II
Merck Index:14,8681