Sodium dihydrogen phosphate dihydrate, BAKER, Laboratory Reagent, J.T.Baker®
Catalog # 1768.1000
Supplier: Avantor
CAS Number:
Specifications
- Pack type:Plastic bottle for solids
- Pk:1 kg
- Seq:4024
Specifications
Specification Test Results
- Assay:min. 98 %
- Arsenic (As):max. 2
Specification Test Results
About this item
:
:156.01 g/mol
Merck Index:12,08806
Boiling Pt:∼650 °C (1013 hPa)
Melting Pt:60 °C
Density:1.91 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-449-2
REACH:01-2119489796-13
CAS Number:13472-35-0
Formula:NaH₂PO₄·2H₂O
MDL Number:MFCD00149209