Order Entry
Austria
ContactUsLinkComponent
Sodium dihydrogen phosphate monohydrate 98.0-103.0% (by anhydrous basis), crystals USP, FCC, J.T.Baker®
Sodium dihydrogen phosphate monohydrate 98.0-103.0% (by anhydrous basis), crystals USP, FCC, J.T.Baker®
Supplier:  Avantor
Certificates
undefined
Sodium dihydrogen phosphate monohydrate 98.0-103.0% (by anhydrous basis), crystals USP, FCC, J.T.Baker®
Supplier:  Avantor
Certificates

Specification Test Results

  • Meets U.S.P Requirements:
    98.0 - 103.0%
  • GMP Manufactured Product:
    Passes Test
  • Meets F.C.C. Requirements:
    ≤3 ppm
  • Food GMP Manufactured Product:
    ≤0.014%
  • CAUTION: For Manufacturing, processing or repackaging:
    Passes Test
  • Bulk Food Chemical:
    Passes Test
  • Bulk Pharmaceutical Chemical:
    ≤0.2%
  • USP - Assay (anhydrous basis):
    98.0 - 103.0 %
  • USP - Aluminum, Calcium, and Related Elements:
    Passes Test
  • USP - Arsenic (As):
    ≤3 ppm
  • USP - Chloride (Cl):
    ≤0.014 %
  • USP - Identification A:
    Passes Test
  • USP - Identification B:
    Passes Test
  • USP - Insoluble Substances:
    ≤0.2 %
  • USP - pH (1 in 20):
    4.1 - 4.5
  • USP - Sulfate (SO₄):
    ≤0.15 %
  • USP - Water (H₂O):
    10.0 - 15.0 %
  • FCC - Assay (anhydrous basis):
    98.0 - 103.0 %
  • FCC - Arsenic (As):
    ≤3 ppm
  • FCC - Identification A:
    Passes Test
  • FCC - Identification B:
    Passes Test
  • FCC - Fluoride (F):
    ≤0.005 %
  • FCC - Insoluble Substances:
    ≤0.2 %
  • FCC - Lead (Pb):
    ≤4 mg/kg
  • FCC - Loss on Drying:
    10.0 - 15.0 %
  • Appearance (White crystals or granules):
    Passes Test

Specification Test Results

Related Information

Frequently Bought Together