Order Entry
Austria
ContactUsLinkComponent
Disodium hydrogen phosphate dodecahydrate, BAKER ANALYZED®, Laboratory Reagent, J.T.Baker®
Disodium hydrogen phosphate dodecahydrate, BAKER ANALYZED®, Laboratory Reagent, J.T.Baker®
Supplier:  Avantor
Certificates
Disodium hydrogen phosphate dodecahydrate, BAKER ANALYZED®, Laboratory Reagent, J.T.Baker®
Supplier:  Avantor
Certificates

Specification Test Results

  • Assay:
    98.0 - 101.0 %
  • Chloride (Cl):
    max. 0.001 %
  • Iron (Fe):
    max. 0.001 %
  • Heavy Metals (as Pb):
    max. 0.001 %
  • Insoluble Matter:
    max. 0.005 %
  • pH of 5% Solution at 25°C:
    8.7 - 9.3
  • Sulfate (SO4):
    max. 0.02 %
  • Nitrogen Compounds (as N):
    max. 5

Specification Test Results

Related Information

Frequently Bought Together