Order Entry
Austria
ContactUsLinkComponent
Trisodium phosphate dodecahydrate, BAKER ANALYZED® ACS, J.T.Baker®
Irritant
Trisodium phosphate dodecahydrate, BAKER ANALYZED® ACS, J.T.Baker®
Catalog # 0307.1000
Supplier:  Avantor
CAS Number:  
Irritant
Trisodium phosphate dodecahydrate, BAKER ANALYZED® ACS, J.T.Baker®
Catalog # 0307.1000
Supplier:  Avantor
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 kg
  • Seq:
    4024

Specifications

Specification Test Results

  • Assay:
    98.0 - 102.0 %
  • Chloride (Cl):
    max. 0.001 %
  • Iron (Fe):
    max. 0.001 %
  • Excess Alkali (as NaOH):
    max. 2.5 %
  • Heavy Metals (as Pb):
    max. 0.001 %
  • Insoluble Matter:
    max. 0.01 %
  • Sulfate (SO4):
    max. 0.01 %

Specification Test Results

About this item

 :
 :380.12 g/mol
Merck Index:13,08736
Melting Pt:∼75 °C
Density:1.62 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-509-8
ADR:8,III
REACH:01-2119489800-32-xxxx
 :GHS07
CAS Number:10101-89-0
Formula:Na₃PO₄·12H₂O
MDL Number:MFCD00149198