Order Entry
Austria
ContactUsLinkComponent
In situ hybridisation wash reagent
In situ hybridisation wash reagent
Catalog # ENZOENZ33809
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
undefined
In situ hybridisation wash reagent
Catalog # ENZOENZ33809
Supplier:  ENZO LIFE SCIENCES
CAS Number:  

Specifications

  • Pk:
    30 mL
  • Description:
    In situ hybridisation wash reagent
  • Seq:
    02

Specifications

About this item

Hybridisation wash reagent provides a high quality hybridisation wash solution for in situ hybridisation analyses using DNA probes labeled with biotin, digoxigenin or fluorescein.

Formulation: Liquid, PBS containing deionised formamide.