Titriplex® IV (trans-1,2-Diaminocyclohexane-N,N,N',N'-tetraacetic acid monohydrate) analytical reagent, Supelco®
Supplier: Merck
Certificates
About this item
CAS Number:125572-95-4
:MFCD00066429
Formula:C₁₄H₂₂N₂O₈·1H₂O
:364.35 g/mol
Melting Pt:213…216 °C
:M-Clarity™ quality level = MQ100
EINECS:236-308-9
:GHS07
Storage Temperature:Ambient
Limited Time Sale
Select items below are currently on sale
Specification Test Results
- Solubility:(20 °C) practically insoluble
- Melting Point:213 °C
- Molar Mass:364.34 g/mol
- Bulk Density:500 kg/m3
Specification Test Results
Related Information
Certificates
Search by Lot NumberMerck Brighter titration Titripur®
Product Literature
msds_108424
SDS
ProductPage_108424
Product specification
Frequently Bought Together