EDTA disodium salt dihydrate, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:6381-92-6
:
Formula:C₁₀H₁₄N₂Na₂O₈·2H₂O
MDL Number:MFCD00150037
:372.24 g/mol
Merck Index:13,03543
Melting Pt:255 °C
:M-Clarity™ quality level = MQ300
Storage Temperature:Ambient
EINECS:205-358-3
Frequently Bought Together