Order Entry
Austria
ContactUsLinkComponent
 
 
 
EDTA disodium salt dihydrate ≥99.0% for HPLC, Fluka™
 
undefined
 
 
EDTA disodium salt dihydrate ≥99.0% for HPLC, Fluka™
 

 

  • Assay:
    99.0 - 101.0 %
  • Suitability test (HPLC):
    complying
  • Colour of the solution:
    complying
  • Clearness of the solution:
    complying
  • Absorbance at 500 nm:
    Max. 0.02
  • Absorbance at 290 nm:
    Max. 0.02
  • Absorbance at 280 nm:
    Max. 0.03
  • Absorbance at 270 nm:
    Max. 0.05
  • Absorbance at 260 nm:
    Max. 0.15
  • Aluminium (Al):
    Max. 5 ppm
  • Barium (Ba):
    Max. 5 ppm
  • Bismuth (Bi):
    Max. 5 ppm
  • Calcium (Ca):
    Max. 10 ppm
  • Cadmium (Cd):
    Max. 5 ppm
  • Cobalt (Co):
    Max. 5 ppm
  • Chromium (Cr):
    Max. 5 ppm
  • Copper (Cu):
    Max. 5 ppm
  • Iron (Fe):
    Max. 5 ppm
  • Potassium (K):
    Max. 50 ppm
  • Lithium (Li):
    Max. 5 ppm
  • Magnesium (Mg):
    Max. 5 ppm
  • Manganese (Mn):
    Max. 5 ppm
  • Molybdenum (Mo):
    Max. 5 ppm
  • Nickel (Ni):
    Max. 5 ppm
  • Lead (Pb):
    Max. 5 ppm
  • Strontium (Sr):
    Max. 5 ppm
  • Zinc (Zn):
    Max. 5 ppm
  • Total S (as SO₄):
    Max. 50 ppm
  • Chloride (Cl):
    Max. 50 ppm

 

Frequently Bought Together