Order Entry
Austria
ContactUsLinkComponent
EDTA disodium salt dihydrate ≥99%, Gen-Apex for molecular biology
EDTA disodium salt dihydrate ≥99%, Gen-Apex for molecular biology
Catalog # 33600.267
Supplier:  VWR Chemicals
CAS Number:  
undefined
EDTA disodium salt dihydrate ≥99%, Gen-Apex for molecular biology
Catalog # 33600.267
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle for solids
  • Pk:
    500 g
  • Seq:
    3645

Specifications

Specification Test Results

  • Assay:
    Min. 99 %
  • Identification (IR):
    Passes test
  • Identification B:
    Passes test Ph.Eur.
  • Identification (Na):
    Passes test
  • Colouration (0,2 mol/l; water):
    Max. 10 APHA
  • Heavy metals (as Pb):
    Max. 5 ppm
  • Transmittance (260 nm) (0.2 mol/l):
    Min. 50 %
  • Transmittance (280 nm) (0.2 mol/l):
    Min. 90 %
  • Transmittance (320 nm) (0.2 mol/l):
    Min. 95 %

Specification Test Results

About this item

 :
 :372.24 g/mol
Merck Index:13,03543
Melting Pt:255 °C
Storage Temperature:Ambient
EINECS:205-358-3
CAS Number:6381-92-6
Formula:C₁₀H₁₄N₂Na₂O₈·2H₂O
MDL Number:MFCD00150037