Order Entry
Austria
ContactUsLinkComponent
 
EDTA disodium salt dihydrate 99.0-101.0% (dried basis), GenAR® USP for biotechnology, Macron Fine Chemicals™
 
undefined
EDTA disodium salt dihydrate 99.0-101.0% (dried basis), GenAR® USP for biotechnology, Macron Fine Chemicals™
 

 

  • GMP Manufactured Product:
  • Meets B.P. Chemical Specifications:
  • Meets E.P. Chemical Specifications:
  • Food GMP Manufactured Product:
  • Meets U.S.P Requirements:
  • Meets F.C.C. Requirements:
  • CAUTION: For Manufacturing, processing or repackaging:
  • Bulk Pharmaceutical Chemical:
  • Bulk Food Chemical:
  • USP - Assay (dried basis):
    99.0 - 101.0 %
  • USP - Identification A:
    Passes Test
  • USP - Identification B:
    Passes Test
  • USP - Identification C:
    Passes Test
  • USP - pH (1 in 20):
    4.0 - 5.5
  • USP - Loss on Drying (by wt):
    8.7 - 11.4 %
  • USP - Calcium (Ca):
    Passes Test
  • USP - Limit of Nitrilotriacetic Acid (by wt):
    ≤0.1 %
  • EP/BP - Appearance of Solution:
    Passes Test
  • EP/BP - Identification A:
    Passes Test
  • EP/BP - Assay:
    98.5 - 101.0 %
  • EP/BP - Identification B:
    Passes Test
  • EP/BP - Identification D:
    Passes Test
  • EP/BP - Impurity A and Related Substances - Impurity A:
    ≤0.1 %
  • EP/BP - Iron (Fe):
    ≤80 ppm
  • EP/BP - pH (1 in 20):
    4.0 - 5.5
  • Endotoxin Concentration (EU/g):
    ≤5
  • FCC - Assay (anhydrous basis):
    99.0 - 101.0 %
  • FCC - Identification A:
    Passes Test
  • FCC - Identification B:
    Passes Test
  • FCC - Identification C:
    Passes Test
  • FCC - Lead (Pb):
    ≤10 mg/kg
  • FCC - pH (1 in 100):
    4.3 - 4.7
  • FCC - Calcium (Ca):
    Passes Test
  • FCC - Limit of Nitrilotriacetic Acid (by wt):
    ≤0.1 %

 

Frequently Bought Together