EDTA disodium salt dihydrate 99.0-101.0%, crystals, BAKER ANALYZED® ACS, J.T.Baker®
: Avantor
:6381-92-6
:
:C₁₀H₁₄N₂Na₂O₈·2H₂O
:MFCD00150037
:372.24 g/mol
:13,03543
:255 °C
:Ambient
:205-358-3
- For Laboratory,Research,or Manufacturing Use:99.0 - 101.0 %
- Assay (C₁₀H₁₄N₂Na₂O₈·2H₂O):99.0 - 101.0 %
- Insoluble Matter:≤0.005 %
- pH of 5% Solution at 25°C:4.0 - 6.0
- Nitrilotriacetic Acid (by polarography):≤0.1 %
- Heavy Metals (as Pb):≤0.005 %
- Iron (Fe):≤0.01 %
Frequently Bought Together