Order Entry
Austria
ContactUsLinkComponent
EDTA disodium salt dihydrate 99.0-101.0%, crystals, Ultrapure Bioreagent for molecular biology, J.T.Baker®
EDTA disodium salt dihydrate 99.0-101.0%, crystals, Ultrapure Bioreagent for molecular biology, J.T.Baker®
Supplier:  Avantor
Certificates
undefined
EDTA disodium salt dihydrate 99.0-101.0%, crystals, Ultrapure Bioreagent for molecular biology, J.T.Baker®
Supplier:  Avantor
Certificates

Specification Test Results

  • For Laboratory,Research,or Manufacturing Use:
    99.0 - 101.0%
  • Assay (C₁₀H₁₄N₂Na₂O₈·2H₂O):
    99.0 - 101.0 %
  • DNase Activity (None Detected):
    Passes Test
  • RNase Activity:
    Passes Test
  • Protease Activity:
    Passes Test
  • pH of 5% Solution at 25°C:
    4.0 - 6.0
  • Calcium (Ca):
    ≤0.005 %
  • Heavy Metals (as Pb):
    ≤0.001 %
  • Insoluble Matter:
    ≤0.005 %
  • Iron (Fe):
    ≤0.005 %
  • Nitrilotriacetic Acid (by polarography):
    ≤0.1 %

Specification Test Results

Related Information

Frequently Bought Together