Order Entry
Austria
ContactUsLinkComponent
 
EDTA disodium salt dihydrate, BAKER ANALYZED®, Laboratory Reagent, J.T.Baker®
 :  Avantor
 
undefined
EDTA disodium salt dihydrate, BAKER ANALYZED®, Laboratory Reagent, J.T.Baker®
 :  Avantor
 

 

  • Assay:
    99.0 - 101.0 %
  • Heavy Metals (as Pb):
    max. 0.001 %
  • Insoluble Matter:
    max. 0.005 %
  • pH of 5% Solution at 25°C:
    4.0 - 6.0
  • Sulfate (SO4):
    max. 0.01 %
  • Trace Impurities (in ppm) Copper (Cu):
    max. 3
  • Trace Impurities (in ppm)Iron (Fe):
    max. 5

 

 

Frequently Bought Together