Order Entry
Austria
ContactUsLinkComponent
EDTA disodium salt dihydrate 99+% for analysis
Irritant
Health
EDTA disodium salt dihydrate 99+% for analysis
Certificates
undefined
Irritant
Health
EDTA disodium salt dihydrate 99+% for analysis
Certificates

Specification Test Results

  • Appearance:
    White Crystalline powder
  • Infrared spectrum:
    Conforms
  • Assay:
    99.0 to 101.0 % (On dry substance)
  • Loss on drying:
    ≤11.4 % (200°C, 3 h)
  • pH:
    4.0 to 6.0 (1 % at 20°C)
  • Iron (Fe):
    ≤80 ppm
  • Heavy metals (as Pb):
    ≤10 ppm

Specification Test Results

Related Information

Frequently Bought Together