Order Entry
Austria
ContactUsLinkComponent
tri-Sodium citrate dihydrate ≥99%, Gen-Apex for molecular biology
tri-Sodium citrate dihydrate ≥99%, Gen-Apex for molecular biology
Catalog # 33615.268
Supplier:  VWR Chemicals
CAS Number:  
undefined
tri-Sodium citrate dihydrate ≥99%, Gen-Apex for molecular biology
Catalog # 33615.268
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle for solids
  • Pk:
    500 g
  • Seq:
    3645

Specifications

Specification Test Results

  • Assay:
    Min. 99 %
  • Colouration (0,1 mol/l; water):
    Max. 10 APHA
  • Heavy metals (as Pb):
    Max. 2 ppm
  • Transmittance (260 nm):
    Min. 98 %
  • Transmittance (280 nm):
    Min. 98 %

Specification Test Results

About this item

 :
 :294.1 g/mol
Merck Index:13,08675
Melting Pt:150 °C
Density:1.76 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:200-675-3
REACH:01-2119457027-40
CAS Number:6132-04-3
Formula:Na₃C₆H₅O₇·2H₂O
MDL Number:MFCD00150031