Order Entry
Austria
ContactUsLinkComponent
 
tri-Sodium citrate dihydrate, powder, EMPROVE® ESSENTIAL Ph. Eur., BP, JP, USP, E331, SAFC®
Catalog # 1.06446.9029
CAS Number:  
undefined
tri-Sodium citrate dihydrate, powder, EMPROVE® ESSENTIAL Ph. Eur., BP, JP, USP, E331, SAFC®
Catalog # 1.06446.9029
CAS Number:  

Specifications

  • Pk:
    25 kg
  • Seq:
    4283

Specifications

About this item

 :
 :294.1 g/mol
Merck Index:13,08675
Melting Pt:150 °C
Density:1.76 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:200-675-3
CAS Number:6132-04-3
Formula:Na₃C₆H₅O₇·2H₂O
MDL Number:MFCD00150031