
Citric acid monohydrate, BAKER ANALYZED® ACS, J.T.Baker®
Supplier: Avantor
Certificates
About this item
CAS Number:5949-29-1
:
Formula:C₆H₈O₇.H₂O
MDL Number:MFCD00149972
:210.14 g/mol
Merck Index:13,02350
Melting Pt:135…152 °C
Density:1.552 g/cm³ (20 °C)
Flash Pt:173.9 °C (closed cup)
EINECS:201-069-1
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Assay:99.0 - 102.0 %
- Chloride (Cl):max. 0.001 %
- Insoluble Matter:max. 0.005 %
- Oxalate (C2O4):passes test
- Phosphate (PO4):max. 0.001 %
- Residue after Ignition:max. 0.02 %
- Sulfur compounds (as SO4):max. 0.002 %
- Substances Carbonizable by Hot H2SO4 (Tartrates, etc.):passes test
- Trace Impurities (in ppm)Iron (Fe):max. 3
- Trace Impurities (in ppm)Lead (Pb):max. 2
Specification Test Results
Related Information
Frequently Bought Together