
di-Ammonium oxalate monohydrate 99.5% for analysis
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:6009-70-7
:
Formula:(NH₄)₂C₂O₄·H₂O
MDL Number:MFCD00149694
:142.11 g/mol
Merck Index:13,00541
Boiling Pt:70 °C (1013 hPa)
Melting Pt:70 °C
Density:~ 1.5 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:214-202-3
ADR:6.1,III
:GHS07
Specification Test Results
- Appearance:White Crystalline powder
- Titration with KMnO4:≥99.4 %
- Chloride (Cl):≤20 ppm
- Sulfate (SO4):≤50 ppm
- Iron (Fe):≤2 ppm
- Heavy metals (as Pb):≤10 ppm
Specification Test Results
Frequently Bought Together