Order Entry
Austria
ContactUsLinkComponent
Zinc acetate dihydrate, BAKER ANALYZED® ACS, J.T.Baker®
Zinc acetate dihydrate, BAKER ANALYZED® ACS, J.T.Baker®
Catalog # 0357.0500
Supplier:  Avantor
CAS Number:  
Zinc acetate dihydrate, BAKER ANALYZED® ACS, J.T.Baker®
Catalog # 0357.0500
Supplier:  Avantor
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    500 g
  • Seq:
    3645
  • Environmentally Preferable:

Specifications

Specification Test Results

  • Assay:
    99.0 - 101.0 %
  • Calcium (Ca):
    max. 0.005 %
  • Potassium (K):
    max. 0.01 %
  • Magnesium (Mg):
    max. 0.005 %
  • Sodium (Na):
    max. 0.05 %
  • Lead (Pb):
    max. 0.002 %
  • Insoluble Matter:
    max. 0.005 %
  • pH of 5% Solution at 25°C:
    6.0 - 7.0
  • Sulfate (SO4):
    max. 0.002 %
  • Chloride (Cl):
    max. 5
  • Iron (Fe):
    max. 5

Specification Test Results

About this item

Synonyms: Acetic acid zinc salt dihydrate, Zinc (II) acetate dihydrate, Zinc diacetate dihydrate
 :
 :219.51 g/mol
Merck Index:12,10256
Melting Pt:237 °C
Density:1.718 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:209-170-2
UN:3077
ADR:9,III
 :GHS07
 :GHS09
CAS Number:5970-45-6
Formula:(H₃CCOO)₂Zn·2H₂O
MDL Number:MFCD00066961