Order Entry
Austria
ContactUsLinkComponent
Sodium acetate trihydrate, BAKER ANALYZED® for HPLC, J.T.Baker®
Sodium acetate trihydrate, BAKER ANALYZED® for HPLC, J.T.Baker®
Catalog # 0393.1000
Supplier:  Avantor
CAS Number:  
undefined
Sodium acetate trihydrate, BAKER ANALYZED® for HPLC, J.T.Baker®
Catalog # 0393.1000
Supplier:  Avantor
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle for solids
  • Pk:
    1 kg
  • Seq:
    4024

Specifications

About this item

 :
 :136.08 g/mol
Merck Index:13,08642
Boiling Pt:120…123 °C (1013 hPa)
Melting Pt:58 °C
Density:1.4 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:204-823-8
CAS Number:6131-90-4
Formula:H₃CCOONa·3H₂O
MDL Number:MFCD00071557