Order Entry
Austria
ContactUsLinkComponent
Sodium acetate trihydrate, BAKER ANALYZED® ACS, J.T.Baker®
Sodium acetate trihydrate, BAKER ANALYZED® ACS, J.T.Baker®
Supplier:  Avantor
Certificates
undefined
Sodium acetate trihydrate, BAKER ANALYZED® ACS, J.T.Baker®
Supplier:  Avantor
Certificates

Specification Test Results

  • Assay:
    99.0 - 101.0 %
  • Calcium (Ca):
    max. 0.005 %
  • Chloride (Cl):
    max. 0.001 %
  • Potassium (K):
    max. 0.005 %
  • Magnesium (Mg):
    max. 0.002 %
  • Insoluble Matter:
    max. 0.005 %
  • pH of 5% Solution at 25°C:
    7.5 - 9.2
  • Substances Reducing KMnO4:
    passes test
  • Sulfate (SO4):
    max. 0.002 %
  • Iron (Fe):
    max. 5
  • Heavy Metals (as Pb):
    max. 5
  • Phosphate (PO4):
    max. 5

Specification Test Results

Related Information

Frequently Bought Together