Order Entry
Austria
ContactUsLinkComponent
Sodium acetate trihydrate 99+%, extra pure
Sodium acetate trihydrate 99+%, extra pure
Certificates
undefined
Sodium acetate trihydrate 99+%, extra pure
Certificates

Specification Test Results

  • Appearance:
    White Adhering crystals
  • Infrared spectrum:
    Conforms
  • Titration with HClO4:
    ≥99.0 %
  • Chloride (Cl):
    ≤200 ppm
  • Sulfate (SO4):
    ≤200 ppm
  • Iron (Fe):
    ≤10 ppm
  • Potassium (K):
    ≤500 ppm
  • Heavy metals (as Pb):
    ≤10 ppm
  • Insoluble matter:
    ≤0.05 %

Specification Test Results

Related Information

Frequently Bought Together