Guanidinium thiocyanate, Millipore®
Catalog # 368975-500
Supplier: Merck Millipore (Calbiochem)
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:500 g
- Seq:3645
- Environmentally Preferable:
Specifications
About this item
Synonyms: Guanidinium sulphocyanide, GTC, Guanidine Thiocyanate, Guanidinium rhodanide
:
:118.16 g/mol
Melting Pt:118 °C
Density:1.29 g/cm³ (20 °C)
EINECS:209-812-1
:GHS07
Storage Temperature:Ambient
CAS Number:593-84-0
Formula:CH₅N₃·CHNS
MDL Number:MFCD00013027