Order Entry
Austria
ContactUsLinkComponent
Guanidinium hydrochloride ≥99%, GPR RECTAPUR®
Irritant
Guanidinium hydrochloride ≥99%, GPR RECTAPUR®
Supplier:  VWR Chemicals
Certificates
undefined
Irritant
Guanidinium hydrochloride ≥99%, GPR RECTAPUR®
Supplier:  VWR Chemicals
Certificates

Specification Test Results

  • Assay:
    Min. 99 %
  • IR Spectrum:
    Passes test
  • Melting point:
    180 - 188 °C

Specification Test Results

Related Information

Frequently Bought Together