Order Entry
Austria
ContactUsLinkComponent
Guanidinium hydrochloride ≥98.5%, Technical Grade low ash residue after ignition
Irritant
Guanidinium hydrochloride ≥98.5%, Technical Grade low ash residue after ignition
Catalog # E424-500G
Supplier:  VWR Chemicals
CAS Number:  
undefined
Irritant
Guanidinium hydrochloride ≥98.5%, Technical Grade low ash residue after ignition
Catalog # E424-500G
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Bottle, amber
  • Pk:
    500 g
  • Seq:
    3645

Specifications

Specification Test Results

  • Ammonium Chloride:
    <0.5%
  • Ash:
    <0.4%
  • Moisture (KF):
    <0.3%
  • Purity:
    >98.5%

Specification Test Results

About this item

 :
 :95.53 g/mol
Merck Index:13,04578
Melting Pt:185 °C
Density:1.354 g/cm³ (20 °C)
EINECS:200-002-3
 :GHS07
Storage Temperature:Ambient
CAS Number:50-01-1
Formula:NH₂C(=NH)NH₂·HCl
MDL Number:MFCD00013026