



Malachite green oxalate
Catalog # MOLE11318220
Supplier: Molekula
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g
- Seq:3308
Specifications
About this item
:
:927.02 g/mol
Merck Index:13,05722
Melting Pt:∼159 °C
Storage Temperature:Ambient
EINECS:219-441-7
UN:2811
ADR:6.1,III
:GHS05
:GHS08
:GHS07
:GHS09
CAS Number:2437-29-8
:18015-76-4
Formula:C₅₂H₅₄N₄O₁₂
MDL Number:MFCD00011766