Order Entry
Austria
ContactUsLinkComponent
 
Corrosive
Health
Irritant
Environment
Malachite green oxalate
Catalog # EHERC14680000
Supplier:  EHRENSTORFER
CAS Number:  
undefined
Corrosive
Health
Irritant
Environment
Malachite green oxalate
Catalog # EHERC14680000
Supplier:  EHRENSTORFER
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    0,25 g
  • Seq:
    3029

Specifications

About this item

 :
 :927.02 g/mol
Merck Index:13,05722
Melting Pt:∼159 °C
Storage Temperature:Ambient
EINECS:219-441-7
UN:2811
ADR:6.1,III
 :GHS05
 :GHS08
 :GHS07
 :GHS09
CAS Number:2437-29-8
 :18015-76-4
Formula:C₅₂H₅₄N₄O₁₂
MDL Number:MFCD00011766