



Malachite green oxalate, pure, certified
:2437-29-8
:18015-76-4
:C₅₂H₅₄N₄O₁₂
:MFCD00011766
:927.02 g/mol
:13,05722
:144…150 °C
:Ambient
:219-441-7
:2811
:6.1,III
:GHS06
:GHS08
:GHS05
:GHS09
- Appearance:Green to dark green Crystalline powder or crystals
- Infrared spectrum:Conforms
- Dye content:≥90 %
- UV Lambda max:614 to 620 nm (in water)
- Absorption Ratio P-15/P+15:1.00 to 1.15
- Visual transition interval:from pH 0.0 (yellow) to pH 2.0 (green)
- BSC Certification:Passes
Frequently Bought Together