N,N,N',N'-Tetramethyl-p-phenylenediamine dihydrochloride ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:637-01-4
:
Formula:C₁₀H₁₈Cl₂N₂
MDL Number:MFCD00012482
:237.17 g/mol
Merck Index:12,09367
Melting Pt:∼ 228 °C (dec.)
EINECS:211-274-8
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together