
N,N-Dimethyl-1,4-phenylenediammonium dichloride 99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:536-46-9
:MFCD00012990
Formula:(CH₃)₂NC₆H₄NH₂·2HCl
MDL Number:MFCD00012991
:209.12 g/mol
Merck Index:13,03280
Melting Pt:222 °C (dec.)
Storage Temperature:Ambient
EINECS:208-635-7
UN:2811
ADR:6.1,II
:GHS06
Specification Test Results
- Appearance:White to yellow to beige to pink to grey Powder
- Infrared spectrum:Conforms
- Solubility:(5 % in water) Clear colorless to yellow to purple-brown
- Titration with HClO4:98.5 to 101.5 %
Specification Test Results
Frequently Bought Together