Order Entry
Austria
ContactUsLinkComponent
TRIS HCl (tris(hydroxymethyl)aminomethane hydrochloride)
TRIS HCl (tris(hydroxymethyl)aminomethane hydrochloride)
Catalog # MOLEM10696487
Supplier:  Molekula
CAS Number:  
undefined
TRIS HCl (tris(hydroxymethyl)aminomethane hydrochloride)
Catalog # MOLEM10696487
Supplier:  Molekula
CAS Number:  

Specifications

  • Pk:
    5 kg
  • Seq:
    4114

Specifications

About this item

 :
 :157.6 g/mol
Merck Index:12,09902
Melting Pt:150…152 °C
Storage Temperature:Ambient
EINECS:214-684-5
CAS Number:1185-53-1
Formula:NH₂C(CH₂OH)₃·HCl
MDL Number:MFCD00012590