Order Entry
Austria
ContactUsLinkComponent
TRIS HCl (tris(hydroxymethyl)aminomethane hydrochloride) ≥99.5%, Proteomics Grade
Irritant
Flammable
TRIS HCl (tris(hydroxymethyl)aminomethane hydrochloride) ≥99.5%, Proteomics Grade
Catalog # M108-1KG
Supplier:  VWR Chemicals
CAS Number:  
undefined
Irritant
Flammable
TRIS HCl (tris(hydroxymethyl)aminomethane hydrochloride) ≥99.5%, Proteomics Grade
Catalog # M108-1KG
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    HDPE bottle
  • Pk:
    1 kg
  • Seq:
    4024

Specifications

Specification Test Results

  • Calcium:
    <0.0001%
  • Copper:
    <0.0001%
  • DNase:
    none detected
  • Electrophoresis:
    Pass
  • Insolubles:
    <0.001%
  • Iron:
    <0.001%
  • Lead:
    <0.0001%
  • Magnesium:
    <0.0001%
  • Manganese:
    <0.0001%
  • pH (0.1 M, Water) @25 °C:
    4.2 – 4.9
  • Protease:
    none detected
  • Purity:
    >99.5
  • RNase:
    none detected
  • Zinc:
    ≤0.0001%

Specification Test Results

About this item

 :
 :157.6 g/mol
Merck Index:12,09902
Melting Pt:150…152 °C
EINECS:214-684-5
 :GHS07
 :GHS02
Storage Temperature:Ambient
CAS Number:1185-53-1
Formula:NH₂C(CH₂OH)₃·HCl
MDL Number:MFCD00012590