Order Entry
Austria
ContactUsLinkComponent
Silver nitrate 0.1 mol/l concentrated aqueous solution, Titrisol®, Supelco®
Corrosive
Environment
Silver nitrate 0.1 mol/l concentrated aqueous solution, Titrisol®, Supelco®
Catalog # 1.09990.0001
Supplier:  Merck
CAS Number:  
undefined
Corrosive
Environment
Silver nitrate 0.1 mol/l concentrated aqueous solution, Titrisol®, Supelco®
Catalog # 1.09990.0001
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Plastic ampoule
  • Pk:
    1 Ampoul
  • Seq:
    0501

Specifications

Specification Test Results

  • Solubility:
    (20 °C) soluble
  • Density:
    1.27 g/cm3 (20 °C)
  • pH:
    7 - 9 (H2O, 20 °C)
  • Hazard Class:
    8
  • Hazard Class Description:
    Corrosive Material

Specification Test Results

About this item

 :
 :169.87 g/mol
ADR:8,
 :M-Clarity™ quality level = MQ100
EINECS:231-853-9
UN:1760
 :GHS05
 :GHS09
Storage Temperature:Ambient
CAS Number:7761-88-8
Formula:AgNO₃
MDL Number:MFCD00003414