Iodine 0.05 mol/l concentrated aqueous solution, Titrisol®, Supelco®
1.09910.0001
: Merck
:
- Pack type:Glass ampoule
- Pk:1 Ampoul
- Seq:0501
- Environmentally Preferable:
- Solubility:(20 °C) soluble
- Density:1.38 g/cm3 (20 °C)
- pH:6.7 (H2O, 20 °C)
:
:253.81 g/mol
:M-Clarity™ quality level = MQ100
:231-442-4
:GHS08
:Ambient
:7553-56-2
:I₂
:MFCD00011355