Order Entry
Austria
ContactUsLinkComponent
Ammonium iron(II) sulphate 0.1 mol/l concentrated aqueous solution, Titrisol®, Supelco®
Ammonium iron(II) sulphate 0.1 mol/l concentrated aqueous solution, Titrisol®, Supelco®
Catalog # 1.09864.0001
Supplier:  Merck
CAS Number:  
Ammonium iron(II) sulphate 0.1 mol/l concentrated aqueous solution, Titrisol®, Supelco®
Catalog # 1.09864.0001
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Plastic ampoule
  • Pk:
    1 Ampoul
  • Seq:
    0501
  • Environmentally Preferable:

Specifications

Specification Test Results

  • Solubility:
    (20 °C) soluble
  • Density:
    1.25 g/cm3 (20 °C)
  • pH:
    0.4 (H2O, 20 °C)
  • Hazard Class:
    8
  • Hazard Class Description:
    Corrosive Material

Specification Test Results

About this item

Synonyms: di-Ammonium iron (II) sulphate, Ammonium ferrous sulphate
 :
 :284.05 g/mol
Melting Pt:100
EINECS:233-151-8
UN:2796
 :GHS05
Storage Temperature:Ambient
CAS Number:10045-89-3
Formula:H₈FeN₂O₈S₂
MDL Number:MFCD00051039
Density:1.86
ADR:8
Merck Index:12,00552
 :M-Clarity™ quality level = MQ100