Order Entry
Austria
ContactUsLinkComponent
Ammonium thiocyanate 0.1 mol/l concentrated aqueous solution, Titrisol®, Supelco®
Ammonium thiocyanate 0.1 mol/l concentrated aqueous solution, Titrisol®, Supelco®
Catalog # 1.09900.0001
Supplier:  Merck
CAS Number:  
Ammonium thiocyanate 0.1 mol/l concentrated aqueous solution, Titrisol®, Supelco®
Catalog # 1.09900.0001
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Plastic ampoule
  • Pk:
    1 Ampoul
  • Seq:
    0501

Specifications

Specification Test Results

  • Solubility:
    (20 °C) soluble
  • Density:
    1.03 g/cm3 (20 °C)
  • pH:
    4.5 (H2O, 20 °C)

Specification Test Results

About this item

 :
 :76.12 g/mol
 :M-Clarity™ quality level = MQ100
Storage Temperature:Ambient
EINECS:217-175-6
CAS Number:1762-95-4
Formula:NH₄SCN
MDL Number:MFCD00011428