

Potassium hydroxide 0.5 mol/l in aqueous solution, Titrisol®, Supelco®
Catalog # 1.09919.0001
Supplier: Merck
CAS Number:
Specifications
- Pack type:Fibre case
- Pk:1 Ampoul
- Seq:0501
Specifications
Specification Test Results
- Solubility:(20 °C) soluble
- Density:1.58 g/cm3 (20 °C)
- pH:13.5 (H2O, 20 °C)
- Hazard Class:8
- Hazard Class Description:Corrosive Material
Specification Test Results
About this item
:
:56.11 g/mol
ADR:8,
:M-Clarity™ quality level = MQ100
EINECS:215-181-3
UN:1814
:GHS05
:GHS07
Storage Temperature:Ambient
CAS Number:1310-58-3
Formula:KOH
MDL Number:MFCD00003553