

Potassium hydroxide concentrate for dilution to 0.1 N in aqueous solution, Acculute™ standard volumetric solution
Catalog # 88604.LS
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pk:6
- Seq:0006
Specifications
About this item
CAS Number:1310-58-3
:
Formula:KOH
MDL Number:MFCD00003553
:56.11 g/mol
Storage Temperature:Ambient
EINECS:215-181-3
UN:1814
ADR:8,II
:GHS05
:GHS07