
Sulphuric acid 0.05 mol/l concentrated aqueous solution, Titrisol®, Supelco®
: Merck
:7664-93-9
:
:H₂SO₄
:MFCD00064589
:98.08 g/mol
:8,
:M-Clarity™ quality level = MQ100
:231-639-5
:2796
:GHS05
:Ambient
- Solubility:(20 °C) soluble
- Density:1.06 g/cm3 (20 °C)
- pH:1 (H2O, 20 °C)
- Hazard Class:8
- Hazard Class Description:Corrosive Material
Frequently Bought Together