Order Entry
Austria
ContactUsLinkComponent
 
Corrosive
Irritant
Hydrochloric acid concentrate for dilution to 2L, 1 M HCl (1N) standard volumetric solution, Fluka™
Catalog # 38281-1EA
CAS Number:  
Corrosive
Irritant
Hydrochloric acid concentrate for dilution to 2L, 1 M HCl (1N) standard volumetric solution, Fluka™
Catalog # 38281-1EA
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    1
  • Seq:
    0001

Specifications

Specification Test Results

  • Factor:
    0.998 - 1.002
  • BAM/EMPA reference material:
    effective
  • BAM/EMPA Certificate Number:
    effective
  • Traceable to NIST SRM:
    effective

Specification Test Results

About this item

 :
Density:1.11 g/cm³
EINECS:231-595-7
ADR:8,II
 :36.46 g/mol
UN:1789
 :GHS05
 :GHS07
Storage Temperature:Ambient
CAS Number:7647-01-0
Formula:HCl
MDL Number:MFCD00011324