

Hydrochloric acid concentrate for dilution to 10L, 1 M HCl (1N) standard volumetric solution, Fluka™
Catalog # 38283-1EA
Supplier: Honeywell Chemicals
CAS Number:
Specifications
- Pack type:Plastic bottle
- Pk:1
- Seq:0001
Specifications
Specification Test Results
- Factor:0.998 - 1.002
- BAM/EMPA reference material:effective
- BAM/EMPA Certificate Number:effective
- Traceable to NIST SRM:effective
Specification Test Results
About this item
:
Density:1.11 g/cm³
EINECS:231-595-7
ADR:8,II
:36.46 g/mol
UN:1789
:GHS05
:GHS07
Storage Temperature:Ambient
CAS Number:7647-01-0
Formula:HCl
MDL Number:MFCD00011324