

Hydrochloric acid concentrate for dilution to 1L, 0.5 M HCl (0.5N) volumetric solution, Fluka™
Supplier: Honeywell Chemicals
Certificates
About this item
CAS Number:7647-01-0
:
Formula:HCl
MDL Number:MFCD00011324
Density:1.11 g/cm³
Storage Temperature:Ambient
EINECS:231-595-7
UN:1789
ADR:8,II
:36.46 g/mol
:GHS07
:GHS05
Specification Test Results
- Factor:0.998 - 1.002
- BAM/EMPA reference material:effective
- BAM/EMPA Certificate Number:effective
- Traceable to NIST SRM:effective
Specification Test Results
Frequently Bought Together