Order Entry
Austria
ContactUsLinkComponent
Hydrochloric acid concentrate for dilution to 0.5 mol in aqueous solution, DILUT-IT®, Laboratory Reagent, J.T.Baker®
Corrosive
Irritant
Hydrochloric acid concentrate for dilution to 0.5 mol in aqueous solution, DILUT-IT®, Laboratory Reagent, J.T.Baker®
Catalog # 4654.
Supplier:  Avantor
CAS Number:  
Corrosive
Irritant
Hydrochloric acid concentrate for dilution to 0.5 mol in aqueous solution, DILUT-IT®, Laboratory Reagent, J.T.Baker®
Catalog # 4654.
Supplier:  Avantor
CAS Number:  

Specifications

  • Pk:
    210 mL
  • Seq:
    2545

Specifications

Specification Test Results

  • Titer (mol/l):
    0.4990-0.5010

Specification Test Results

About this item

 :
ADR:8,II
 :36.46 g/mol
EINECS:231-595-7
UN:1789
 :GHS05
 :GHS07
Storage Temperature:Ambient
CAS Number:7647-01-0
Formula:HCl
MDL Number:MFCD00011324