Order Entry
Austria
ContactUsLinkComponent
Titriplex® III concentrate 0.1 M, Titrisol®, Supelco®
Health
Titriplex® III concentrate 0.1 M, Titrisol®, Supelco®
Catalog # 1.09992.0001
Supplier:  Merck
CAS Number:  
Health
Titriplex® III concentrate 0.1 M, Titrisol®, Supelco®
Catalog # 1.09992.0001
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Plastic ampoule
  • Pk:
    1 Ampoul
  • Seq:
    0501

Specifications

Specification Test Results

  • Solubility:
    (20 °C) soluble
  • Density:
    1.14 g/cm3 (20 °C)
  • pH:
    7.4 - 7.6 (H2O, 20 °C)

Specification Test Results

About this item

 :
 :336.21 g/mol
Density:1.14 g/cm3 (20 °C)
 :M-Clarity™ quality level = MQ100
EINECS:205-358-3
 :GHS08
Storage Temperature:Ambient
CAS Number:139-33-3
Formula:C₁₀H₁₄N₂Na₂O₈
MDL Number:MFCD00070672