ContactUsLinkComponent
L(+)-Cysteine hydrochloride monohydrate, TECHNICAL
  23255.186
 :  
undefined
L(+)-Cysteine hydrochloride monohydrate, TECHNICAL
  23255.186
 :  
  • Pack type:
    Plastic bottle for solids
  • Pk:
    100 g
  • Seq:
    3401
  • Identification:
    Passes test
 :
 :175.64 g/mol
 :13,02810
 :168…170 °C
 :Ambient
 :200-157-7
 :7048-04-6
 :HSCH₂CH(NH₂)COOH·HCl·H₂O
 :MFCD00065606