Order Entry
Austria
Orders LinkContactUsLinkComponent
Buffer, TBE solution 10X concentrate (TRIS-borate-EDTA buffer)

Buffer, TBE solution 10X concentrate (TRIS-borate-EDTA buffer)

TBE buffer solution 10X concentrate (TRIS-borate-EDTA buffer), Glass bottle, 1 L
Catalog # MOBO15002-1000
Marketsource
Supplier:   MO BIO
Buffer, TBE solution 10X concentrate (TRIS-borate-EDTA buffer)
Catalog # MOBO15002-1000
Supplier:  MO BIO
Marketsource
CAS Number:  
Buffer, TBE solution 10X concentrate (TRIS-borate-EDTA buffer)

TBE buffer solution 10X concentrate (TRIS-borate-EDTA buffer), Glass bottle, 1 L

CAS Number:  
Price
Inquire For Price
Log in for availability
1 * 1 L
Product Details & Documents
Specifications

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 L
  • Description:
    TBE buffer solution 10X concentrate (TRIS-borate-EDTA buffer)

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.