Order Entry
Austria
ContactUsLinkComponent
Buffer, TBE solution 10X concentrate (TRIS-borate-EDTA buffer), Millipore®
Buffer, TBE solution 10X concentrate (TRIS-borate-EDTA buffer), Millipore®
Catalog # 574795-1
CAS Number:  
Buffer, TBE solution 10X concentrate (TRIS-borate-EDTA buffer), Millipore®
Catalog # 574795-1
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 L
  • Description:
    TBE buffer solution 10X concentrate (TRIS-borate-EDTA buffer)

Specifications

About this item