Order Entry
Austria
ContactUsLinkComponent
Buffer, TAE solution 50X concentrate (TRIS-acetate-EDTA buffer)
Buffer, TAE solution 50X concentrate (TRIS-acetate-EDTA buffer)
Catalog # B49
CAS Number:  
undefined
Buffer, TAE solution 50X concentrate (TRIS-acetate-EDTA buffer)
Catalog # B49
CAS Number:  

Specifications

  • Pk:
    1 KIT
  • Description:
    TAE buffer solution 50X concentrate (TRIS-acetate-EDTA buffer)

Specifications

About this item