Order Entry
Austria
ContactUsLinkComponent
Ethidium bromide 10 mg/ml in aqueous solution for electrophoresis, Sigma-Aldrich®
Ethidium bromide 10 mg/ml in aqueous solution for electrophoresis, Sigma-Aldrich®
Catalog # 1.11608.0030
Supplier:  Merck
CAS Number:  
Ethidium bromide 10 mg/ml in aqueous solution for electrophoresis, Sigma-Aldrich®
Catalog # 1.11608.0030
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    30 mL
  • Seq:
    2345

Specifications

About this item

 :
ADR:6.1,
 :M-Clarity™ quality level = MQ100
 :394.31 g/mol
EINECS:214-984-6
UN:2810
 :GHS08
 :GHS07
CAS Number:1239-45-8
Formula:C₂₁H₂₀BrN₃
MDL Number:MFCD00011724